2-Bromoquinoline-4-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR906018-1G OR906018-250MG
APOSOR906018-1GEA 202 EUR
APOSOR906018-1G APOSOR906018-250MG
2-Bromoquinoline-4-carboxylic acid 97%
2-Bromoquinoline-4-carboxylic acid
Formel: C₁₀H₆BrNO₂
Molecular Weight: 252,0600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08705669
CAS-Nummer: 15733-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR