6-Chloroquinazolin-4-amine 98%

Lieferant: Apollo Scientific

OR906153-1G OR906153-250MG
APOSOR906153-1GEA 210 EUR
APOSOR906153-1G APOSOR906153-250MG
6-Chloroquinazolin-4-amine 98%
6-Chloroquinazolin-4-amine
Formel: C₈H₆ClN₃
Schmelzpunkt: 300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02676502
CAS-Nummer: 19808-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR