Methyl-2-(2-aminophenyl)acetate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-(2-aminophenyl)acetate
OR906172-1G
OR906172-250MG
APOSOR906172-1GEA
143
EUR
APOSOR906172-1G
APOSOR906172-250MG
Methyl-2-(2-aminophenyl)acetate 95%
Methyl-2-(2-aminophenyl)acetate
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973395 CAS-Nummer: 35613-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...