Methyl-3-(cyclopentylamino)propanoate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-(cyclopentylamino)propanoate
OR906174-1G
OR906174-5G
APOSOR906174-1GEA
62.5
EUR
APOSOR906174-1G
APOSOR906174-5G
Methyl-3-(cyclopentylamino)propanoate 95%
Methyl-3-(cyclopentylamino)propanoate
Formel:
C₉H₁₇NO₂ Molecular Weight: 171,2368 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10687312 CAS-Nummer: 754125-43-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...