1,7-Dihydroxynaphthalin 95%

Lieferant: Apollo Scientific

Synonyme: 1,7-Naphthalindiol

OR906184-25G
APOSOR906184-25GEA 88.6 EUR
APOSOR906184-25G
1,7-Dihydroxynaphthalin 95%
1,7-Dihydroxynaphthalin
Formel: C₁₀H₈O₂
Molecular Weight: 160,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00035720
CAS-Nummer: 575-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR