2-[Methyl(phenyl)amino]ethanol 98%

Lieferant: Apollo Scientific

Synonyme: N-Methyl-N-2-ethanolaniline, 2-(Methyl(phenyl)amino)ethanol, N-(2-Hydroxyethyl)-N-methylaniline

OR906200-500G
APOSOR906200-500GEA 115 EUR
APOSOR906200-500G
2-[Methyl(phenyl)amino]ethanol 98%
2-(N-Methylanilino)ethanol
Formel: C₉H₁₃NO
Molecular Weight: 151,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020572
CAS-Nummer: 93-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR