Ethyl-2-(thiazol-2-yl)acetate 95%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-(thiazol-2-yl)acetate
OR906252-1G
OR906252-250MG
APOSOR906252-1GEA
201
EUR
APOSOR906252-1G
APOSOR906252-250MG
Ethyl-2-(thiazol-2-yl)acetate 95%
Ethyl-2-(thiazol-2-yl)acetate
Formel:
C₇H₉NO₂S Molecular Weight: 171.2200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12114489 CAS-Nummer: 141704-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...