6-Nitro-1H-indole-2-carboxylic acid 95%

Lieferant: Apollo Scientific

Synonyme: 6-Nitroindole-2-carboxylic acid

OR906264-1G OR906264-5G
APOSOR906264-1GEA 142 EUR
APOSOR906264-1G APOSOR906264-5G
6-Nitro-1H-indole-2-carboxylic acid 95%
6-Nitro-1H-indole-2-carboxylic acid
Formel: C₉H₆N₂O₄
Schmelzpunkt: 304…305 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02664466
CAS-Nummer: 10242-00-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR