2-Methoxy-6-methylbenzonitril 95%
Lieferant: Apollo Scientific
OR906288-1G
OR906288-5G
APOSOR906288-1GEA
31.9
EUR
APOSOR906288-1G
APOSOR906288-5G
2-Methoxy-6-methylbenzonitril 95%
2-Methoxy-6-methylbenzonitril
Formel:
C₉H₉NO Molecular Weight: 147,1739 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10699539 CAS-Nummer: 53005-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...