1-Chlor-3-methoxypropan 98%

Lieferant: Apollo Scientific

Synonyme: 3-Chloropropyl methyll ether

OR906289-100G OR906289-25G
APOSOR906289-100GEA 502 EUR
APOSOR906289-100G APOSOR906289-25G
1-Chlor-3-methoxypropan 98%
1-Chloro-3-methoxypropane
Formel: C₄H₉ClO
Molecular Weight: 108.57 g/mol
Dichte: 0.999
Lagertemperatur: Raumtemperatur
MDL: MFCD05663419
CAS-Nummer: 36215-07-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR