Methyl quinoline-3-carboxylate 95%

Lieferant: Apollo Scientific

OR906465-1G OR906465-5G
APOSOR906465-1GEA 119 EUR
APOSOR906465-1G APOSOR906465-5G
Methyl quinoline-3-carboxylate 95%
Methyl quinoline-3-carboxylate
Formel: C₁₁H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03550859
CAS-Nummer: 53951-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR