N-Boc-propargylamin 98%
Lieferant: Apollo Scientific
Synonyme:
N-Boc-propargylamine, 2-Propynylcarbamic Acid tert-Butyl Ester, tert-Butyl 2-Propynylcarbamate
OR906557-100G
APOSOR906557-100GEA
154
EUR
APOSOR906557-100G
N-Boc-propargylamin 98%
N-Boc-propargylamin
Formel:
C₈H₁₃NO₂ Molecular Weight: 155,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07367245 CAS-Nummer: 92136-39-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...