1-Isocyano-4-methoxybenzene 95%

Lieferant: Apollo Scientific

Synonyme: 4-Methoxyphenyl isocyanide

OR906675-1G OR906675-5G
APOSOR906675-1GEA 197 EUR
APOSOR906675-1G APOSOR906675-5G
1-Isocyano-4-methoxybenzene 95%
1-Isocyano-4-methoxybenzene
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02664592
CAS-Nummer: 10349-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR