2-Amino-4-morpholin-4-ylpyrimidine 95%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-4-morpholin-4-yl-pyrimidine
OR906697-1G
OR906697-250MG
APOSOR906697-1GEA
106
EUR
APOSOR906697-1G
APOSOR906697-250MG
2-Amino-4-morpholin-4-ylpyrimidine 95%
2-Amino-4-morpholin-4-ylpyrimidine
Formel:
C₈H₁₂N₄O Molecular Weight: 180,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11520888 CAS-Nummer: 861031-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...