(S)-methyl-2-aminobutanoate 96%

Lieferant: Apollo Scientific

Synonyme: H-Abu-OMe·HCl

OR906728-1G OR906728-5G
APOSOR906728-1GEA 261 EUR
APOSOR906728-1G APOSOR906728-5G
(S)-methyl-2-aminobutanoate 96%
(S)-methyl-2-aminobutanoate
Formel: C₅H₁₁NO₂
Molecular Weight: 117.148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09037399
CAS-Nummer: 15399-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR