Ethyl-2-oxocyclohexancarboxylat 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-oxocyclohexanecarboxylate

OR906774-100G OR906774-500G
APOSOR906774-100GEA 72.5 EUR
APOSOR906774-100G APOSOR906774-500G
Ethyl-2-oxocyclohexancarboxylat 95%
Ethyl-2-oxocyclohexancarboxylat
Formel: C₉H₁₄O₃
Molecular Weight: 170,21 g/mol
Dichte: 1,067 g/cm³ (25 °C)
Flammpunkt: 96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001631
CAS-Nummer: 1655-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR