5-Chloropyrimidin-4-ol 95%

Lieferant: Apollo Scientific

OR906830-1G OR906830-250MG OR906830-5G
APOSOR906830-1GEA 328 EUR
APOSOR906830-1G APOSOR906830-250MG APOSOR906830-5G
5-Chloropyrimidin-4-ol 95%
5-Chloropyrimidin-4-ol
Formel: C4H3ClN2O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 4349-08-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR