1-Methylpyrrol-3-carbonsäure 97%

Lieferant: Apollo Scientific

Synonyme: 1-Methylpyrrole-3-carboxylic Acid

OR906844-5G
APOSOR906844-5GEA 253 EUR
APOSOR906844-5G
1-Methylpyrrol-3-carbonsäure 97%
1-Methylpyrrole-3-carboxylic acid
Formel: C₆H₇NO₂
Molecular Weight: 125.1300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111733
CAS-Nummer: 36929-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR