3-(Piperidin-1-ylmethyl)phenylboronsäure 95%
Lieferant: Apollo Scientific
OR906854-1G
OR906854-250MG
OR906854-5G
APOSOR906854-1GEA
265
EUR
APOSOR906854-1G
APOSOR906854-250MG
APOSOR906854-5G
3-(Piperidin-1-ylmethyl)phenylboronsäure 95%
3-(Piperidin-1-ylmethyl)phenylboronsäure
Formel:
C₁₂H₁₈BNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08060620 CAS-Nummer: 1032610-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...