Methyl-3-brompropionat 97%

Lieferant: Apollo Scientific

Synonyme: Methyl 3-bromopropanoate

OR906916-500G
APOSOR906916-500GEA 114 EUR
APOSOR906916-500G
Methyl-3-brompropionat 97%
Methyl-3-brompropionat
Formel: C₄H₇BrO₂
Molecular Weight: 167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000250
CAS-Nummer: 3395-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR