Salicylaldehydethylether 98%
Lieferant: Apollo Scientific
Synonyme:
2-Ethoxybenzaldehyde
OR906957-100G
APOSOR906957-100GEA
58
EUR
APOSOR906957-100G
Salicylaldehydethylether 98%
Salicylaldehydethylether
Formel:
C₉H₁₀O₂ Molecular Weight: 150,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003316 CAS-Nummer: 613-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...