cis-4-Cyclohexen-1,2-dicarbonsäure 95%
Lieferant: Apollo Scientific
Synonyme:
(1R,2S)-rel-Cyclohex-4-ene-1,2-dicarboxylic acid
OR906968-500G
APOSOR906968-500GEA
102
EUR
APOSOR906968-500G
cis-4-Cyclohexen-1,2-dicarbonsäure 95%
cis-4-Cyclohexen-1,2-dicarbonsäure
Formel:
C₈H₁₀O₄ Molecular Weight: 170,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070482 CAS-Nummer: 2305-26-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...