3'-Chlor-4'-hydroxyacetanilid 98%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-4-hydroxyacetanilide, N-(3-Chloro-4-hydroxyphenyl)acetamide

OR906989-10G OR906989-1G OR906989-5G
APOSOR906989-10GEA 230 EUR
APOSOR906989-10G APOSOR906989-1G APOSOR906989-5G
3'-Chlor-4'-hydroxyacetanilid 98%
3'-Chlor-4'-hydroxyacetanilid
Formel: C₈H₈ClNO₂
Molecular Weight: 185.6100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01318123
CAS-Nummer: 3964-54-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR