4-Methoxyoxindole 95%
Lieferant: Apollo Scientific
Synonyme:
4-Methoxyindolin-2-one, 4-Methoxy-2-indolinone
OR906993-1G
OR906993-250MG
APOSOR906993-1GEA
168
EUR
APOSOR906993-1G
APOSOR906993-250MG
4-Methoxyoxindole 95%
4-Methoxyoxindole
Formel:
C₉H₉NO₂ Molecular Weight: 163,1733 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368221 CAS-Nummer: 7699-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...