Aminoacetamide 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Glycinamide

OR907030-25G OR907030-5G
APOSOR907030-25GEA 654 EUR
APOSOR907030-25G APOSOR907030-5G
Aminoacetamide 98%
Aminoacetamide
Formel: C₂H₆N₂O
Molecular Weight: 74,083 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01859715
CAS-Nummer: 598-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR