2-Amino-3-chlor-5-methylpyrazin 95%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-5-methylpyrazin-2-amine

OR907053-100MG OR907053-1G OR907053-250MG
APOSOR907053-100MGEA 71.2 EUR
APOSOR907053-100MG APOSOR907053-1G APOSOR907053-250MG
2-Amino-3-chlor-5-methylpyrazin 95%
2-Amino-3-chloro-5-methylpyrazine
Formel: C₅H₆ClN₃
Molecular Weight: 143,5700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834805
CAS-Nummer: 89182-14-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR