5-Hydroxy-2H-benzo[b][1,4]oxazin-3(4H)-one 96%
Lieferant: Apollo Scientific
OR907081-1G
OR907081-250MG
APOSOR907081-1GEA
531
EUR
APOSOR907081-1G
APOSOR907081-250MG
5-Hydroxy-2H-benzo[b][1,4]oxazin-3(4H)-one 96%
5-Hydroxy-2H-benzo[b][1,4]oxazin-3(4H)-one
Formel:
C8H7NO3 Lagertemperatur: Raumtemperatur |
CAS-Nummer:
177210-33-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...