Methyl-2-(4-formylphenoxy)acetate 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-(4-formylphenoxy)acetate
OR907093-1G
OR907093-5G
APOSOR907093-1GEA
46.4
EUR
APOSOR907093-1G
APOSOR907093-5G
Methyl-2-(4-formylphenoxy)acetate 95%
Methyl-2-(4-formylphenoxy)acetate
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194,180 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00463122 CAS-Nummer: 73620-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...