N-Methoxy-N-methylbenzamide 98%

Lieferant: Apollo Scientific

APOSOR907094-25GEA 109 EUR
APOSOR907094-25G APOSOR907094-5G APOSOR907094-100G
N-Methoxy-N-methylbenzamide 98%
N-Methoxy-N-methylbenzamide
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00075320
CAS-Nummer: 6919-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR