4-Bromo-N-methylpyrimidin-2-amine 98%
Lieferant: Apollo Scientific
OR907174-1G
OR907174-250MG
APOSOR907174-1GEA
43.5
EUR
APOSOR907174-1G
APOSOR907174-250MG
4-Bromo-N-methylpyrimidin-2-amine 98%
4-Bromo-N-methylpyrimidin-2-amine
Formel:
C₅H₆BrN₃ Molecular Weight: 188,0300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13193672 CAS-Nummer: 1209458-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...