4-Bromo-2,6-dimethylbenzoic acid

Lieferant: Apollo Scientific

OR907248-25G OR907248-5G
APOSOR907248-25GEA 395 EUR
APOSOR907248-25G APOSOR907248-5G
4-Bromo-2,6-dimethylbenzoic acid
4-Bromo-2,6-dimethylbenzoic acid
Formel: C₉H₉BrO₂
Molecular Weight: 229,0706 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09965997
CAS-Nummer: 74346-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR