4-Brom-5-chloranthranilsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-4-bromo-5-chlorobenzoic acid

OR907297-1G OR907297-25G OR907297-5G
APOSOR907297-1GEA 21.9 EUR
APOSOR907297-1G APOSOR907297-25G APOSOR907297-5G
4-Brom-5-chloranthranilsäure 98%
4-Brom-5-chloranthranilsäure
Formel: C₇H₅BrClNO₂
Molecular Weight: 250,4800 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10000554
CAS-Nummer: 150812-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR