N-Ethyl-3-nitropyridin-4-amine 95%
Lieferant: Apollo Scientific
Synonyme:
4-(Ethylamino)-3-nitropyridine
OR907331-1G
OR907331-250MG
APOSOR907331-1GEA
125
EUR
APOSOR907331-1G
APOSOR907331-250MG
N-Ethyl-3-nitropyridin-4-amine 95%
N-Ethyl-3-nitropyridin-4-amine
Formel:
C₇H₉N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02266948 CAS-Nummer: 562825-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...