3-Phenylpiperazin-2-one 95%

Lieferant: Apollo Scientific

OR907410-1G OR907410-250MG
APOSOR907410-1GEA 97.3 EUR
APOSOR907410-1G APOSOR907410-250MG
3-Phenylpiperazin-2-one 95%
3-Phenylpiperazin-2-one
Formel: C₁₀H₁₂N₂O
Molecular Weight: 176,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871361
CAS-Nummer: 5368-28-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR