2-Ethoxyisonicotinsäure 98%
Lieferant: Apollo Scientific
Synonyme:
2-Ethoxypyridine-4-carboxylic acid
OR907519-25G
APOSOR907519-25GEA
626
EUR
APOSOR907519-25G
2-Ethoxyisonicotinsäure 98%
2-Ethoxyisonicotinsäure
Formel:
C₈H₉NO₃ Molecular Weight: 167,1620 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115719 CAS-Nummer: 91940-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...