5-Methoxy-2-pyridinecarboxylic acid 98%

Lieferant: Apollo Scientific

Synonyme: 5-Methoxypicolinic acid

OR907539-10G OR907539-1G OR907539-5G
APOSOR907539-10GEA 272 EUR
APOSOR907539-10G APOSOR907539-1G APOSOR907539-5G
5-Methoxy-2-pyridinecarboxylic acid 98%
5-Methoxy-2-pyridinecarboxylic acid
Formel: C7H7NO3
Molecular Weight: 153,1400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06800961
CAS-Nummer: 29082-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR