4-Chlor-N,N-dimethylbenzamid 98%

Lieferant: Apollo Scientific

Synonyme: N,N-Dimethyl 4-chlorobenzamide

OR907561-25G OR907561-5G
APOSOR907561-25GEA 473 EUR
APOSOR907561-25G APOSOR907561-5G
4-Chlor-N,N-dimethylbenzamid 98%
4-Chlor-N,N-dimethylbenzamid
Formel: C₉H₁₀ClNO
Molecular Weight: 183.6400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463859
CAS-Nummer: 14062-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR