Ethyl-2-(3-bromophenyl)acetate 97%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 2-(3-bromophenyl)acetate
OR907639-25G
APOSOR907639-25GEA
78.3
EUR
APOSOR907639-25G
Ethyl-2-(3-bromophenyl)acetate 97%
Ethyl-2-(3-bromophenyl)acetate
Formel:
C₁₀H₁₁BrO₂ Molecular Weight: 243,1000 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09953130 CAS-Nummer: 14062-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...