3-Methoxy-2-naphthaldehyd 98%
Lieferant: Apollo Scientific
OR907835-5G
APOSOR907835-5GEA
112
EUR
APOSOR907835-5G
3-Methoxy-2-naphthaldehyd 98%
3-Methoxy-2-naphthaldehyd
Formel:
C₁₂H₁₀O₂ Molecular Weight: 186,2066 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13191799 CAS-Nummer: 56679-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...