1-Cyclopentylethanone 95%

Lieferant: Apollo Scientific

OR907909-25G OR907909-100G
APOSOR907909-25GEA 69.6 EUR
APOSOR907909-25G APOSOR907909-100G
1-Cyclopentylethanone 95%
1-Cyclopentylethanone
Formel: C₇H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00060799
CAS-Nummer: 6004-60-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR