2-Cyano-2-Methylpropionsäure Ethylester 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Ethyl 2-Cyano-2-methylpropionate, 2-Cyano-2-methylpropionic Acid Ethyl Ester
OR908044-100G
OR908044-25G
APOSOR908044-100GEA
253
EUR
APOSOR908044-100G
APOSOR908044-25G
2-Cyano-2-Methylpropionsäure Ethylester 98%
Ethyl-2-cyano-2-methylpropionate
Formel:
C₇H₁₁NO₂ Molecular Weight: 141,17 g/mol Siedepunkt: 185 °C Dichte: 0,97 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00034667 CAS-Nummer: 1572-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...