Methyl-2-cyano-2-methylpropanoate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-cyano-2-methylpropanoate, Methyl 2-cyano-2,2-dimethylacetate

OR908083-100G
APOSOR908083-100GEA 116 EUR
APOSOR908083-100G
Methyl-2-cyano-2-methylpropanoate 95+%
Methyl-2-cyano-2-methylpropanoate
Formel: C₆H₉NO₂
Molecular Weight: 127,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03428328
CAS-Nummer: 72291-30-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR