4-Chlorphenylcyclopropylketon 96%

Lieferant: Apollo Scientific

Synonyme: (4-Chlorophenyl)(cyclopropyl)methanone

OR908214-100G OR908214-25G
APOSOR908214-100GEA 330 EUR
APOSOR908214-100G APOSOR908214-25G
4-Chlorphenylcyclopropylketon 96%
4-Chlorphenylcyclopropylketon
Formel: C₁₀H₉ClO
Molecular Weight: 180.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00001295
CAS-Nummer: 6640-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR