4-(2-Methoxyethoxy)benzeneboronic acid 97%

Lieferant: Apollo Scientific

Synonyme: (4-(2-Methoxyethoxy)phenyl)boronic acid

OR908296-10G OR908296-5G
APOSOR908296-10GEA 116 EUR
APOSOR908296-10G APOSOR908296-5G
4-(2-Methoxyethoxy)benzeneboronic acid 97%
4-(2-Methoxyethoxy)benzeneboronic acid
Formel: C₉H₁₃BO₄
Molecular Weight: 196.009 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03452784
CAS-Nummer: 265664-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR