3,4-Dichlor-o-toluidin 95%
Lieferant: Apollo Scientific
Synonyme:
3,4-Dichloro-2-methylaniline
OR908621-10G
OR908621-1G
OR908621-5G
APOSOR908621-10GEA
754
EUR
APOSOR908621-10G
APOSOR908621-1G
APOSOR908621-5G
3,4-Dichlor-o-toluidin 95%
3,4-Dichlor-o-toluidin
Formel:
C₇H₇Cl₂N Lagertemperatur: Raumtemperatur |
MDL:
MFCD11855814 CAS-Nummer: 62077-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...