2-Brom-4-iodtoluol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-iodo-1-methylbenzene, 1-Bromo-5-iodo-2-metnylbenzene
OR909079-100G
APOSOR909079-100GEA
165
EUR
APOSOR909079-100G
2-Brom-4-iodtoluol 98%
2-Brom-4-iodtoluol
Formel:
C₇H₆BrI Molecular Weight: 296,9300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07779014 CAS-Nummer: 26670-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...