6-Bromo-3,4-dihydro-2H-1,4-benzoxazine
Lieferant: Apollo Scientific
Synonyme:
6-Bromo-3,4-dihydro-2H-benzo[b][1,4]oxazine
OR909084-25G
OR909084-5G
APOSOR909084-25GEA
478
EUR
APOSOR909084-25G
APOSOR909084-5G
6-Bromo-3,4-dihydro-2H-1,4-benzoxazine
6-Bromo-3,4-dihydro-2H-1,4-benzoxazine
Formel:
C₈H₈BrNO Molecular Weight: 214,062 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08544341 CAS-Nummer: 105655-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...