2-Chlor-4-thiocyanatoanilin 97%

Lieferant: Apollo Scientific

OR909137-10G OR909137-1G OR909137-5G
APOSOR909137-10GEA 574 EUR
APOSOR909137-10G APOSOR909137-1G APOSOR909137-5G
2-Chlor-4-thiocyanatoanilin 97%
2-Chlor-4-thiocyanatoanilin
Formel: C₇H₅ClN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD04614735
CAS-Nummer: 3226-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR