3-Chlor-2-methyl-4-thiocyanatoanilin 98%

Lieferant: Apollo Scientific

OR909145-1G OR909145-5G
APOSOR909145-1GEA 168 EUR
APOSOR909145-1G APOSOR909145-5G
3-Chlor-2-methyl-4-thiocyanatoanilin 98%
3-Chlor-2-methyl-4-thiocyanatoanilin
Formel: C₈H₇ClN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11855819
CAS-Nummer: 14030-84-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR