7,8-Dichlorchinolin 98%
Lieferant: Apollo Scientific
OR909167-1G
OR909167-5G
APOSOR909167-1GEA
219
EUR
APOSOR909167-1G
APOSOR909167-5G
7,8-Dichlorchinolin 98%
7,8-Dichloroquinoline
Formel:
C₉H₅Cl₂N Lagertemperatur: Raumtemperatur |
MDL:
MFCD12026044 CAS-Nummer: 703-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...